missing translation for 'onlineSavingsMsg'
Learn More

Benzyltriphenylphosphonium Chloride, TRC

Product Code. 30338268
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30338268 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30338268 Supplier Toronto Research Chemicals Supplier No. TRCB31616010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifica

Chemical Name or Material Benzyltriphenylphosphonium Chloride
CAS 1100-88-5
Molecular Formula C25H22P.Cl
Quantity 10 g
Synonym BPP, BTPPC, GM 200, NSC 116712, Triphenylbenzylphosphonium Chloride
InChI Formula InChI=1S/C25H22P.ClH/c1-5-13-22(14-6-1)21-26(23-15-7-2-8-16-23,24-17-9-3-10-18-24)25-19-11-4-12-20-25;/h1-20H,21H2;1H/q+1;/p-1
SMILES [Cl-].C(c1ccccc1)[P+](c2ccccc2)(c3ccccc3)c4ccccc4
IUPAC Name benzyl(triphenyl)phosphanium;chloride
Molecular Weight (g/mol) 388.9
Formula Weight 388.1148
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.