missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Berberine chloride hydrate

Product Code. 16469877
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16469877 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16469877 Supplier MedChemExpress Supplier No. HY17577
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Berberine chloride hydrate (Natural Yellow 18 chloride hydrate) is an alkaloid that acts as an antibiotic. Berberine chloride hydrate induces reactive oxygen species (ROS) generation and inhibits DNA topoisomerase. Antineoplastic properties.

Specifications

Content And Storage 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Molecular Weight (g/mol) 389.83 g/mol
Shipping Condition Room temperature
For Use With (Application) Cancer-programmed cell death
CAS 68030-18-2
Product Type Natural Products
Purity 98.0%
Solubility DMSO : ≥ 3.9 mg/mL (10.00 mM) ∣H2O : 1 mg/mL (2.57 mM∣ultrasonic and warming and heat to 60°C)
Synonym Natural Yellow 18 chloride hydrate
Target Topoisomerase∣Autophagy∣Bacterial∣Reactive Oxygen Species∣Antibiotic
Quantity 5 g
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.