missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Bergapten

Product Code. 16482519
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16482519 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16482519 Supplier MedChemExpress Supplier No. HYN0370
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Bergapten is a natural anti-inflammatory and anti-tumor agent. Bergapten is inhibitory towards mouse and human CYP isoforms.

Specifications

Content And Storage Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Molecular Weight (g/mol) 216.19 g/mol
Shipping Condition Room temperature
For Use With (Application) Cancer-programmed cell death
CAS 484-20-8
Product Type Natural Products
Purity 99.77%
Solubility DMSO : 20 mg/mL (92.51 mM∣Need ultrasonic) ∣H2O : 0.1 mg/mL (0.46 mM∣Need ultrasonic)
Synonym 5-Methoxypsoralen
Target Cytochrome P450∣Autophagy
Quantity 1 g
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.