missing translation for 'onlineSavingsMsg'
Learn More

Berkshire™ BCR™ Mop 4 (Large Flat Mop Frame)

Product Code. 17856914 Shop All Berkshire Products
Change view
Click to view available options
Autoclavable:
Autoclavable
Unit Size:
Each
Product Code. Autoclavable unitSize
17856914 Autoclavable Each
1 options
This item is not returnable. View return policy

Product Code. 17856914

Brand: Berkshire™ BCR.MOP4.1

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Berkshire BCR™ Mop 4 (Large Flat Mop Frame) is light and easy to maneuver, even with wet wipers attached

  • Clean, light and durable
  • High quality anodized aluminum Clean, light and durable
  • High quality anodized aluminum
  • Autoclavable
  • Flexible system with a wide range of alternative Mop Wipers

Specifications

Color Gray
Sterility Non-sterile
Autoclavable Autoclavable
Size 3.5 x 23 in. (9 x 58 cm)
Quantity 1
For Use With (Application) ISO Class 3 to 7, Floors, Walls and Ceilings
Material Aluminum
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.