missing translation for 'onlineSavingsMsg'
Learn More

Berkshire™ Sterile SatPax™ 1000 Presaturated Wipers

Product Code. 18284589 Shop All Berkshire Products
Change view
Click to view available options
Dimensions (L x W):
9 x 9 in. (22.86 x 22.86 cm)
Unit Size:
Pack of 24
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
18284589 9 x 9 in. (22.86 x 22.86 cm) Pack of 24
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18284589 Supplier Berkshire™ Supplier No. SSP100000124
  • ONLINE EXCLUSIVE PRICE
    602.00 €
    572.39 € / Pack of 24
    Save 29.61 €
    4% Off
View all promotions
 Restricted Item
Estimated Shipment: 19-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Certificate of Sterility provided with each case.

For use in ISO Class 5 (Class 100) applications

  • Made of a nonwoven blend of 55% cellulose and 45% polyester that has been sterilized with gamma radiation to a Sterility Assurance Level of 10-6
  • Presaturated with 70% isopropyl alcohol/30% DI water for one-step disinfecting or cleaning
  • A lot number on each pack assures individual lot quality and traceability, and sterility assurance

Order Info

Basis weight: 67.6g/m2. Packed 30 C-folded sheets per resealable flexible pouch.

Compliance

Bioburden testing and sterility dose audit have been performed, meeting ANSI/ISO/AAMI American National Standards.

Specifications

Product Type Pre-saturated Wipe
Length (English) 9 in.
Length (Metric) 22.86 cm
Width (English) 9 in.
Width (Metric) 22.86 cm
Material Polyester Cellulose
Basis Weight 67.6
Certifications/Compliance RoHS 2002/05, REACH
Dimensions (L x W) 9 x 9 in. (22.86 x 22.86 cm)
For Use With (Application) For use in ISO Class 5 and higher sterile cleanroom environments, final cleaning of surfaces or products prior to manufacturing or packaging
Saturation Solution 70% Isopropyl Alcohol, 30% Deionised Water
Sterility Sterile
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Sterile SatPax 1000
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: isopropanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.