missing translation for 'onlineSavingsMsg'
Learn More

Beryllium sulfate tetrahydrate, 99.99% (metals basis), Thermo Scientific Chemicals

Product Code. 11307158
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11307158 100 g 100g
11397148 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11307158 Supplier Thermo Scientific Alfa Aesar Supplier No. 016104.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

In Industries it is used for beryllium salt making, ceramics and chemical reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7787-56-6
Molecular Formula BeH8O8S
Molecular Weight (g/mol) 177.128
MDL Number MFCD00149156
InChI Key DIMYTQPLZWDZFE-UHFFFAOYSA-L
Synonym beryllium sulfate tetrahydrate, ccris 85, beryllium sulphate tetrahydrate, beryllium monosulfate tetrahydrate, unii-2tyk3lf8zn, beryllium sulfate, tetrahydrate 1:1:4, 2tyk3lf8zn, sulfuric acid, beryllium salt 1:1 , tetrahydrate, beryllium sulfate tetrahydrate beryllium and beryllium compounds, dsstox_cid_4603
PubChem CID 62672
ChEBI CHEBI:53502
IUPAC Name beryllium;sulfate;tetrahydrate
SMILES [Be+2].O.O.O.O.[O-]S(=O)(=O)[O-]

Specifications

Physical Form Lump
Quantity 100 g
UN Number UN1566
Merck Index 14,1175
Solubility Information Freely soluble in water; Insoluble in alcohol
Formula Weight 177.14 (105.07 Anhydrous)
Percent Purity 99.99%
Density 1.1713 g/mL
Assay (metals basis)
Chemical Name or Material Beryllium sulfate tetrahydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.