missing translation for 'onlineSavingsMsg'
Learn More

Beryllium Sulfate Tetrahydrate, Purum p.a., ≥99.0% (T), Honeywell™

Product Code. 15601410 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Poly bottle
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15601410 25 g Poly bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15601410 Supplier Honeywell Research Chemicals Supplier No. 1427025G
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Melting Point 550°C to 600°C
CAS 7787-56-6
Color White
Physical Form Crystalline
Quantity 25 g
Molecular Formula BeH8O8S
Linear Formula BeSO4 · 4H2O
MDL Number MFCD00149156
UN Number UN1566
Synonym beryllium sulfate tetrahydrate, ccris 85, beryllium sulphate tetrahydrate, beryllium monosulfate tetrahydrate, unii-2tyk3lf8zn, beryllium sulfate, tetrahydrate 1:1:4, 2tyk3lf8zn, sulfuric acid, beryllium salt 1:1 , tetrahydrate, beryllium sulfate tetrahydrate beryllium and beryllium compounds, dsstox_cid_4603
InChI Key DIMYTQPLZWDZFE-UHFFFAOYSA-L
SMILES [Be+2].O.O.O.O.[O-]S(=O)(=O)[O-]
IUPAC Name beryllium;sulfate;tetrahydrate
Molecular Weight (g/mol) 177.128
PubChem CID 62672
ChEBI CHEBI:53502
Formula Weight 177.14g/mol
Percent Purity ≥99.0% (T)
Grade purum p.a.
Packaging Poly bottle
Density 1.713g/mL at 25°C
Chemical Name or Material Beryllium Sulfate Tetrahydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.