missing translation for 'onlineSavingsMsg'
Learn More

BES, 99%

Product Code. 11493107
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11493107 100 g 100g
11483107 25 g 25g
11403117 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11493107 Supplier Thermo Scientific Alfa Aesar Supplier No. A16092.22
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 10191-18-1 | C6H15NO5S | 213.25 g/mol

BES is used as a buffer to maintain the pH of the solutions in biological experiments. It is useful for diagnostic assay manufacturing industry. It induces cross-linking between sulfonated polyimide chains in sulfonated polyimide membranes. It acts as biobuffer to investigate the aqueous medium self-assembly of heterometallic CuII/Li 3D coordination polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water.

Notes
Store in cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10191-18-1
Molecular Formula C6H15NO5S
Molecular Weight (g/mol) 213.25
MDL Number MFCD00007533
InChI Key AJTVSSFTXWNIRG-UHFFFAOYSA-N
Synonym n,n-bis 2-hydroxyethyl-2-aminoethanesulfonic acid, bes, ethanesulfonic acid, 2-bis 2-hydroxyethyl amino, 2-bis 2-hydroxyethyl amino ethanesulfonic acid, n,n-bis 2-hydroxyethyl taurine, n,n-bis hydroxyethyl-2-aminoethanesulfonic acid, bes buffering agent, taurine, n,n-bis 2-hydroxyethyl, 2-bis 2-hydroxyethyl amino-ethanesulfonicacid
PubChem CID 73243
ChEBI CHEBI:39045
IUPAC Name 2-[bis(2-hydroxyethyl)amino]ethanesulfonic acid
SMILES OCCN(CCO)CCS(O)(=O)=O

Specifications

Melting Point 153°C to 157°C
Quantity 100 g
Beilstein 1781572
Solubility Information Soluble in water.
Formula Weight 213.25
Percent Purity 99%
Chemical Name or Material BES

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.