missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals BES sodium salt, 99%

Product Code. 11925291 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11925291 25 g 25g
11955291 100 g 100g
11935291 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11925291 Supplier Thermo Scientific Chemicals Supplier No. 439030250
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 66992-27-6 ∣ C6H14NO5S·Na ∣ 235.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 66992-27-6
Molecular Formula C6H14NO5S·Na
Molecular Weight (g/mol) 235.24
InChI Key CFQLQLSIZOWFNV-UHFFFAOYSA-M
Synonym bes sodium salt, sodium 2-bis 2-hydroxyethyl amino ethanesulfonate, n,n-bis 2-hydroxyethyl-2-aminoethanesulfonic acid sodium salt, sodium 2-bis 2-hydroxyethyl amino ethane-1-sulfonate, ethanesulfonic acid, 2-bis 2-hydroxyethyl amino-, monosodium salt, 2-bis 2-hydroxyethyl amino ethanesulfonic acid, sodium salt, bes na, bes sodium salt buffer, potassium c6h14no5s
PubChem CID 23671640
IUPAC Name sodium;2-[bis(2-hydroxyethyl)amino]ethanesulfonate
SMILES C(CO)N(CCO)CCS(=O)(=O)[O-].[Na+]

Specifica

Melting Point 155.6°C to 156.5°C
Color White
CAS Min % 98.5
CAS Max % 100.0
Physical Form Crystalline Powder
Quantity 25 g
Infrared Spectrum Authentic
Packaging Glass Bottle
Assay Percent Range 99%
Formula Weight 235.24
Percent Purity 99%
Grade Ungraded
Chemical Name or Material BES sodium salt
Vedi altri risultati Mostra meno risultati
Identificativo prodotto
  • BES sodium salt

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.