missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals beta-Apo-oxytetracycline, 'can be used as secondary standard'

Product Code. 10104920 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 mg
Packaging:
Glass bottle
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10104920 100 mg Glass bottle 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10104920 Supplier Thermo Scientific Chemicals Supplier No. 257731000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Beta-apo-oxytetracycline, CAS # 18751-99-0, is a member of the tetracycline family and is a degradation production formed from oxytetracycline. This compound displays antibiotic activity.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Information

• Beta-apo-oxytetracycline is a degradation product derived from oxytetracycline and displays antibiotic activity
• Similar to its parent compound, oxytetracycline, beta-apo-oxytetracycline can also inhibit protein synthesis by binding to the 30S ribosomal subunit

Applications

• In laboratory assays, it promotes the inhibition of the growth of aerobic sludge bacteria, Pseudomonas, Agrobacterium, Moraxella, Bacillus, as well as tetracycline-resistant strains of E. coli

TRUSTED_SUSTAINABILITY

Specifications

CAS 18751-99-0
Melting Point 200.0°C to 204.0°C
Color Brown to Brown-Yellow
Infrared Spectrum Authentic
Assay Percent Range 80% min. (as is,calculated)
Packaging Glass bottle
Molecular Formula C22H22N2O8
MDL Number MFCD00062827
Quantity 100 mg
Merck Index 10, 6846
Specific Rotation − 207.00
Synonym beta-apo-oxytetracycline, alpha-apo-oxytetracycline, alpha-apooxytetracycline, alpha-apoterramycin, beta-apo oxytetracycline, 2z-2-amino hydroxy methylidene-5-4,5-dihydroxy-9-methyl-3-oxo-1h-benzo f 2 benzofuran-1-yl-4-dimethylamino-6-hydroxycyclohexane-1,3-dione, 4-1,3-dihydro-4,5-dihydroxy-9-methyl-3-oxonaphtho 2,3-c furan-1-yl-3-dimethylamino-2,5-dihydroxy-6-oxo-1-cyclohexene-1-carboxamide, 4-4,5-dihydroxy-9-methyl-3-oxo-1,3-dihydronaphtho 2,3-c furan-1-yl-3-dimethylamino-2,5-dihydroxy-6-oxocyclohex-1-ene-1-carboxamide, 4-4,5-dihydroxy-9-methyl-3-oxohydrobenzo f isobenzofuranyl-3-dimethylamino-2,5-dihydroxy-6-oxocyclohex-1-enecarboxamide, 4-4,5-dihydroxy-9-methyl-3-oxo-1h-naphtho 2,3-c furan-1-yl-3-dimethylamino-2,5-dihydroxy-6-oxocyclohex-1-ene-1-carboxamide
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alkaline solution and ammonia
InChI Key DRKMHDAKULCOKQ-UHFFFAOYSA-N
SMILES CC1=C2C(OC(=O)C2=C(C3=C1C=CC=C3O)O)C4C(C(=C(C(=O)C4O)C(=O)N)O)N(C)C
IUPAC Name 4-(4,5-dihydroxy-9-methyl-3-oxo-1H-benzo[f][2]benzofuran-1-yl)-3-(dimethylamino)-2,5-dihydroxy-6-oxocyclohexene-1-carboxamide
Molecular Weight (g/mol) 442.39
PubChem CID 54710410
Formula Weight 442.39
Percent Purity ≥97.0% (anhydrous and solvent free), ≥80 wt% (as is, calculated)(HPLC)
Physical Form Powder
Chemical Name or Material β-Apo-oxytetracycline
Show More Show Less
compliance-icons
Product Identifier
  • ß-Apo-oxytetracycline, ''can be used as secondary standard''
Signal Word
  • Warning
Hazard Category
  • Reproductive toxicity Category 2
Hazard Statement
  • H361d-Suspected of damaging the unborn child.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.