missing translation for 'onlineSavingsMsg'
Learn More

Beta-Carotene, TRC

Product Code. 17988594
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17988594 25 mg 25mg
17919374 10 mg 10mg
17918334 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17988594 Supplier Toronto Research Chemicals Supplier No. TRCC18425025MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7235-40-7
Molecular Formula C40 H56
Molecular Weight (g/mol) 536.87
Synonym β-Carotene, all-trans- (8CI), (all-E)-1,1'-(3,7,12,16-Tetramethyl-1,3,5,7,9,11,13,15,17-octadecanonaene-1,18-diyl)bis[2,6,6-trimethylcyclohexene], BetaVit, Betacarotene, C.I. 40800, C.I. Food Orange 5, Carofertin, Carotaben, Caroten Base 35468, Carotene Base 80, Carotene Base 80S, Carotene Base 80SV, CoroCare, Cyclohexene, 1,1'-(3,7,12,16-tetramethyl-1,3,5,7,9,11,13,15,17-octadecanonaene-1,18-diyl)bis[2,6,6-trimethyl-, (all-E)-, Food Orange 5, KPMK, Lucaratin, Lucarotin, Lucarotin 10CWD/O, Lucarotin 20MCT, Lucarotin 30SUN, Lurotin, NSC 62794, Provatene, Provatenol, Rovimix β-carotene, Serlabo, Solatene, all-E-β-Carotene, all-trans-β-Carotene, β-Carotene
IUPAC Name 1,3,3-trimethyl-2-[(1E,3E,5E,7E,9E,11E,13E,15E,17E)-3,7,12,16-tetramethyl-18-(2,6,6-trimethylcyclohexen-1-yl)octadeca-1,3,5,7,9,11,13,15,17-nonaenyl]cyclohexene
SMILES C\C(=C/C=C/C=C(\C)/C=C/C=C(\C)/C=C/C1=C(C)CCCC1(C)C)\C=C\C=C(/C)\C=C\C2=C(C)CCCC2(C)C

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C40H56/c1-31(19-13-21-33(3)25-27-37-35(5)23-15-29-39(37,7)8)17-11-12-18-32(2)20-14-22-34(4)26-28-38-36(6)24-16-30-40(38,9)10/h11-14,17-22,25-28H,15-16,23-24,29-30H2,1-10H3/b12-11+,19-13+,20-14+,27-25+,28-26+,31-17+,32-18+,33-21+,34-22+
Formula Weight 536.4382
Chemical Name or Material beta-Carotene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.