missing translation for 'onlineSavingsMsg'
Learn More

beta-Caryophyllene Epoxide, TRC

Product Code. 30011153
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011153 10 g 10g
30006086 25 g 25g
17903005 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011153 Supplier Toronto Research Chemicals Supplier No. TRCC18473010G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1139-30-6
Molecular Formula C15 H24 O
Molecular Weight (g/mol) 220.35
Synonym 5-Oxatricyclo[8.2.0.04,6]dodecane, 4,12,12-trimethyl-9-methylene-, (1R,4R,6R,10S)-, 5-Oxatricyclo[8.2.0.04,6]dodecane, 4,12,12-trimethyl-9-methylene-, [1R-(1R*,4R*,6R*,10S*)]-, Caryophyllene oxide (6CI), Caryophyllene, epoxide (7CI), (-)-Caryophyllene oxide, (-)-Epoxydihydrocaryophyllene, (-)-β-Caryophyllene epoxide, (-)-β-Caryophyllene oxide, (1R,4R,6R,10S)-4,12,12-Trimethyl-9-methylene-5-oxatricyclo[8.2.0.04,6]dodecane, 4β,5α-Epoxycaryophyllene, 6,7-Epoxy-3(15)-caryophyllene, Caryophyllene 4β,5α-epoxide, Caryophyllene 4β,5α-oxide, trans-Caryophyllene oxide, β-Caryophyllene epoxide, β-Caryophyllene oxide
IUPAC Name (1R,4R,6R,10S)-4,12,12-trimethyl-9-methylidene-5-oxatricyclo[8.2.0.04,6]dodecane
SMILES CC1(C)C[C@H]2[C@H]1CC[C@@]3(C)O[C@@H]3CCC2=C

Specifications

Quantity 10 g
InChI Formula InChI=1S/C15H24O/c1-10-5-6-13-15(4,16-13)8-7-12-11(10)9-14(12,2)3/h11-13H,1,5-9H2,2-4H3/t11-,12-,13-,15-/m1/s1
Formula Weight 220.1827
Chemical Name or Material Caryophyllene Oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.