missing translation for 'onlineSavingsMsg'
Learn More

beta-Cyclocitral, 90+%

Product Code. 11481100
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11481100 10 g 10g
11491100 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11481100 Supplier Thermo Scientific Alfa Aesar Supplier No. B25700.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 432-25-7 | C10H16O | 152.24 g/mol

beta-Cyclocitral is used as flavoring agent, nutrient, stabilizers, surfactants and emulsifiers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
beta-Cyclocitral is used as flavoring agent, nutrient, stabilizers, surfactants and emulsifiers.

Solubility
Soluble in chloroform, dichloromethane, ethyl acetate and methanol. Insoluble in water.

Notes
Air sensitive. Store away from air, oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 432-25-7
Molecular Formula C10H16O
Molecular Weight (g/mol) 152.24
MDL Number MFCD00079078
InChI Key MOQGCGNUWBPGTQ-UHFFFAOYSA-N
Synonym beta-cyclocitral, b-cyclocitral, 1-cyclohexene-1-carboxaldehyde, 2,6,6-trimethyl, .beta.-cyclocitral, 2,6,6-trimethyl-1-cyclohexene-1-carboxaldehyde, unii-77y0u2x29g, cyclocitral, alpha beta-cyclocitral, 2,6,6-trimethylcyclohex-1-ene-1-carbaldehyde, 1-formyl-2,6,6-trimethyl-1-cyclohexene
PubChem CID 9895
ChEBI CHEBI:53177
IUPAC Name 2,6,6-trimethylcyclohexene-1-carbaldehyde
SMILES CC1=C(C=O)C(C)(C)CCC1

Specifications

Density 0.944
Boiling Point 62°C to 63°C (3 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.498
Quantity 10 g
Sensitivity Air Sensitive
Solubility Information Soluble in chloroform,dichloromethane,ethyl acetate and methanol. Insoluble in water.
Formula Weight 152.23
Percent Purity ≥90%
Chemical Name or Material beta-Cyclocitral

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.