missing translation for 'onlineSavingsMsg'
Learn More

beta-Cyclodextrin hydrate

Product Code. 11471888
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11471888 5 g 5g
11481888 25 g 25g
11491888 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11471888 Supplier Thermo Scientific Alfa Aesar Supplier No. A14529.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 68168-23-0 | C42H70O35 | 1134.99 g/mol

beta-Cyclodextrin hydrate forms clathrates. Produces a water-soluble complex with dansyl chloride for the fluorescent labeling of proteins. It is used in pharmaceutical research, as pharmaceutical intermediates and also as chemical reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
beta-Cyclodextrin hydrate forms clathrates. Produces a water-soluble complex with dansyl chloride for the fluorescent labeling of proteins. It is used in pharmaceutical research, as pharmaceutical intermediates and also as chemical reagents.

Solubility
Slightly soluble in water (18.5 g/L at 25°C).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68168-23-0
Molecular Formula C42H70O35
Molecular Weight (g/mol) 1134.99
MDL Number MFCD00150811
InChI Key WHGYBXFWUBPSRW-UHFFFAOYSA-N
Synonym beta-cyclodextrin hydrate, schardinger, a-dextrin hydrate, beta-cyclodextrin hydrate, puriss hplc
PubChem CID 53486154
IUPAC Name 5,10,15,20,25,30,35-heptakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29,32,34-tetradecaoxaoctacyclo[31.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶.2²⁸,³¹]nonatetracontane-36,37,38,39,40,41,42,43,44,45,46,47,48,49-tetradecol
SMILES OCC1OC2OC3C(CO)OC(OC4C(CO)OC(OC5C(CO)OC(OC6C(CO)OC(OC7C(CO)OC(OC8C(CO)OC(OC1C(O)C2O)C(O)C8O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O

Spécification

Melting Point 288°C (decomposition)
Quantity 5 g
Beilstein 5915513
Merck Index 14,2718
Solubility Information Slightly soluble in water (18.5g/L at 25°C).
Optical Rotation +142° (c=1.5 in Water)
Formula Weight 1134.98 (Anhydrous)
Chemical Name or Material beta-Cyclodextrin hydrate

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.