missing translation for 'onlineSavingsMsg'
Learn More

beta-Cyclodextrin Sulfobutyl Ethers, Sodium Salts, TRC

Product Code. 30358631
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30358631 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30358631 Supplier Toronto Research Chemicals Supplier No. TRCC9878451G
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Betadex Sulfobutyl Ether Sodium
CAS 182410-00-0
Molecular Formula C42H70O35•xNa•x(C4H9O3S)
Quantity 1 g
Synonym Betadex Sulfobutyl Ether Sodium Salt
InChI Formula InChI=1S/C42H70O35.C4H10O3S.Na/c43-1-8-29-15(50)22(57)36(64-8)72-30-9(2-44)66-38(24(59)17(30)52)74-32-11(4-46)68-40(26(61)19(32)54)76-34-13(6-48)70-42(28(63)21(34)56)77-35-14(7-49)69-41(27(62)20(35)55)75-33-12(5-47)67-39(25(60)18(33)53)73-31-10(3-45)65-37(71-29)23(58)16(31)51;1-2-3-4-5-6-7-8;/h8-63H,1-7H2;8H,2-4H2,1H3;/t8-,9-,10-,11-,12-,13-,14-,15-,16-,17-,18?,19?,20-,21-,22-,23-,24-,25-,26-,27-,28-,29-,30-,31-,32-,33-,34-,35-,36-,37-,38-,39-,40+,41-,42-;;/m1../s1
SMILES O[C@@H]([C@@H]1O[C@H]([C@@H](C([C@@H]2O[C@H]([C@@H]([C@H]([C@@H]3O[C@H]4O[C@@H]([C@H]([C@@H]([C@H]4O)O)O5)CO)O)O)O[C@@H]3CO)O)O)O[C@@H]2CO)[C@H]([C@H](O[C@@H]1CO)O[C@H]6[C@@H]([C@H]([C@H](O[C@@H]6CO)O[C@H]7[C@@H]([C@H]([C@H](O[C@@H]7CO)O[C@H]8C([C@H]([C@H]5O[C@@H]8CO)O)O)O)O)O)O)O
IUPAC Name β-Cyclodextrin, sulfobutyl ethers, sodium salts
Molecular Weight (g/mol) 1451.3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.