missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals beta-Endorphin, human

Product Code. 15444769
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15444769 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15444769 Supplier Thermo Scientific Chemicals Supplier No. J62374.M^
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

3465.06 g/mol

beta-endorphins produce analgesia by binding to opioid receptors (particularly of the mu subtype) at both pre- and post- synaptic nerve terminals, primarily exerting their effect through presynaptic binding. In the central nervous system, beta-endorphins similarly bind mu-opioid receptors and exert their primary action at presynaptic nerve terminals. It is a potent pain suppressant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Species Human
Purification Method HPLC
Format Powder
For Use With (Application) An endogenous opioid peptide neurotransmitter found in the neurons of both the central and peripheral nervous systemy
Molecular Weight (g/mol) 3465.06
Product Type beta-Endorphin, human
Quantity 1 mg
Regulatory Status RUO - research use only

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.