missing translation for 'onlineSavingsMsg'
Learn More

Beta-Estradiol Precipitable by Digitonin. MP Biomedicals

Product Code. 11498020 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11498020 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11498020 Supplier MP Biomedicals Supplier No. 0210165601
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 50-28-2
Molecular Formula C18H24O2
Quantity 1 g
Synonym estradiol, beta-estradiol, 17beta-estradiol, oestradiol, dihydrofolliculin, estrace, ovocyclin, progynon, dihydrotheelin, dihydroxyestrin
InChI Key VOXZDWNPVJITMN-ZBRFXRBCSA-N
SMILES CC12CCC3C(C1CCC2O)CCC4=C3C=CC(=C4)O
IUPAC Name (8R,9S,13S,14S,17S)-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthrene-3,17-diol
Molecular Weight (g/mol) 272.388
PubChem CID 5757
ChEBI CHEBI:16469
Formula Weight 272.4
Chemical Name or Material Beta-Estradiol, Precipitable by Digitonin.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.