missing translation for 'onlineSavingsMsg'
Learn More

beta-Glycerol Phosphate Disodium Salt Pentahydrate, TRC

Product Code. 30135464
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
Unit Size:
100g
10g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135464 100 g 100g
30008261 10 g 10g
30009650 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135464 Supplier Toronto Research Chemicals Supplier No. TRCG598450100G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13408-09-8
Molecular Formula C3 H7 O6 P . 2 Na
Molecular Weight (g/mol) 216.04
Synonym 1,2,3-Propanetriol, 2-(dihydrogen phosphate), sodium salt, hydrate (1:2:5) (ACI), 1,2,3-Propanetriol, 2-(dihydrogen phosphate), disodium salt, pentahydrate (9CI), Glycerol, 2-(dihydrogen phosphate), disodium salt, pentahydrate (8CI), Sodium β-glycerophosphate pentahydrate, Disodium 2-glycerophosphate pentahydrate, Disodium β-glycerin phosphate pentahydrate, Disodium β-glycerophosphate pentahydrate, Glycerol-2-phosphate disodium salt pentahydrate, β-Glycerophosphate disodium salt pentahydrate
IUPAC Name disodium;1,3-dihydroxypropan-2-yl phosphate
SMILES [Na+].[Na+].OCC(CO)OP(=O)([O-])[O-]

Specifications

Quantity 100 g
InChI Formula InChI=1S/C3H9O6P.2Na/c4-1-3(2-5)9-10(6,7)8;;/h3-5H,1-2H2,(H2,6,7,8);;/q;2*+1/p-2
Formula Weight 215.9776
Chemical Name or Material beta-Glycerol Phosphate Disodium Salt Pentahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.