missing translation for 'onlineSavingsMsg'
Learn More

Beta-Mangostin, TRC

Product Code. 30476422
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
Unit Size:
1mg
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30476422 1 mg 1mg
30472892 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30476422 Supplier Toronto Research Chemicals Supplier No. TRCM1638951MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20931-37-7
Molecular Formula C25 H28 O6
Molecular Weight (g/mol) 424.49
Synonym 1,6-Dihydroxy-3,7-dimethoxy-2,8-bis(3-methyl-2-butenyl)-9H-xanthen-9-one
IUPAC Name 1,6-dihydroxy-3,7-dimethoxy-2,8-bis(3-methylbut-2-enyl)xanthen-9-one
SMILES COc1cc2Oc3cc(O)c(OC)c(CC=C(C)C)c3C(=O)c2c(O)c1CC=C(C)C

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C25H28O6/c1-13(2)7-9-15-18(29-5)12-20-22(23(15)27)24(28)21-16(10-8-14(3)4)25(30-6)17(26)11-19(21)31-20/h7-8,11-12,26-27H,9-10H2,1-6H3
Formula Weight 424.1886
Chemical Name or Material beta-Mangostin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.