missing translation for 'onlineSavingsMsg'
Learn More

Beta Max™-Esliquid Scintillation Cocktail, MP Biomedicals™

Product Code. 15792242 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
4 x 1 gal.
Unit Size:
15.14 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15792242 4 x 1 gal. 15.14 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15792242 Supplier MP Biomedicals Supplier No. 0188002004
 Restricted Item
Estimated Shipment: 29-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Features high tritium efficiency and is excellent with dry filters and test wipes. MP Biomedicals Beta Max™-Esliquid Scintillation Cocktail is available in a large, 5 gallon option or in four 1 gallon containers.

  • Biodegradable, non-toxic, non-flammable and odorless
  • Product can be applied with organic samples, dry filters, test wipes and 3H Acetyl CoA CAT assay extractions
  • Store at room temperature (15–30°C)

Specifications

Boiling Point 300°C
Density 0.9 - 1.0g/mL at 20°C
Quantity 4 x 1 gal.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.