missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals beta-Methyl-DL-phenylalanine hydrochloride, 99%

Product Code. 10215194 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10215194 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10215194 Supplier Thermo Scientific Chemicals Supplier No. 202660010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 80997-87-1 | C10H14ClNO2 | 215.68 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 80997-87-1
CAS Min % 98.5
CAS Max % 100.0
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 99%
Molecular Formula C10H14ClNO2
Packaging Glass Bottle
Linear Formula C6H5CH(CH3)CH(NH2)CO2H·HCl
MDL Number MFCD00012623
Synonym 2-amino-3-phenylbutanoic acid hydrochloride, apba, 2-amino-3-phenylbutanoic acid, chloride, beta-methyl-3-phenyl-dl-alanine hcl, beta-methyl-3-phenyl-dl-alanine hydrochloride, acmc-20ao59, beta-methyl dl phenylalanine hydrochloride, beta-methyl-dl-phenylalanine hydrochloride, 2-azanyl-3-phenyl-butanoic acid hydrochloride, phenylalanine, b-methyl-, hydrochloride 1:1
InChI Key SGKWQBMDOXGYAA-UHFFFAOYNA-N
SMILES Cl.CC(C(N)C(O)=O)C1=CC=CC=C1
IUPAC Name 2-amino-3-phenylbutanoic acid;hydrochloride
Molecular Weight (g/mol) 215.68
Quantity 1 g
PubChem CID 12743546
Formula Weight 215.68
Percent Purity ≥98.5%
Physical Form Powder
Chemical Name or Material β-Methyl-DL-phenylalanine hydrochloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.