missing translation for 'onlineSavingsMsg'
Learn More

beta-NADP monosodium salt, Thermo Scientific™

Product Code. 15805198 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Plastic bottle
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15805198 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15805198 Supplier Thermo Scientific Supplier No. J22653.22
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 1184-16-3
Physical Form Powder
Quantity 100 g
Molecular Formula C21H27N7NaO17P3
MDL Number MFCD00036973
Synonym nadp sodium salt, beta-nicotinamide adenine dinucleotide phosphate sodium
InChI Key JNUMDLCHLVUHFS-QYZPTAICSA-M
SMILES [Na+].NC(=O)C1=CC=C[N+](=C1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]2O[C@H]([C@H](OP(O)(O)=O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O
IUPAC Name [[(2R,3R,4R,5R)-5-(6-aminopurin-9-yl)-3-hydroxy-4-phosphonooxyoxolan-2-yl]methoxy-hydroxyphosphoryl] [(2R,3S,4R,5R)-5-(3-carbamoylpyridin-1-ium-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate;sodium
Molecular Weight (g/mol) 765.39
PubChem CID 131845262
Formula Weight 837.45
Packaging Plastic bottle
Chemical Name or Material beta-NADP monosodium salt
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.