missing translation for 'onlineSavingsMsg'
Learn More

Beta-NADPH Tetrasodium Salt Hydrate, TRC

Product Code. 30136660
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136660 500 mg 500mg
30132502 1 g 1g
17958224 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136660 Supplier Toronto Research Chemicals Supplier No. TRCN201500500MG
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 2412169-80-1
Molecular Formula C21 H26 N7 O17 P3 . 4 Na . x H2 O
Quantity 500 mg
Synonym 2'-(Dihydrogen Phosphate) 5'-(Trihydrogen Pyrophosphate)adenosine 5'5'-Ester 1,4-Dihydro-1-β-D-ribofuranosylnicotinamide Tetrasodium Salt Hydrate, β-NADPH, 2′-NADPH Hydrate, Coenzyme II Reduced Tetrasodium Salt Hydrate, Triphosphopyridine Nucleotide Reduced Tetrasodium Salt Hydrate, Dihydronicotinamide Adenine Dinucleotide Phosphate Tetrasodium Salt Hydrate
InChI Formula InChI=1S/C21H30N7O17P3.4Na.H2O/c22-17-12-19(25-7-24-17)28(8-26-12)21-16(44-46(33,34)35)14(30)11(43-21)6-41-48(38,39)45-47(36,37)40-5-10-13(29)15(31)20(42-10)27-3-1-2-9(4-27)18(23)32;;;;;/h1,3-4,7-8,10-11,13-16,20-21,29-31H,2,5-6H2,(H2,23,32)(H,36,37)(H,38,39)(H2,22,24,25)(H2,33,34,35);;;;;1H2/q;4*+1;/p-4/t10-,11-,13-,14-,15-,16-,20-,21-;;;;;/m1...../s1
SMILES O.[Na+].[Na+].[Na+].[Na+].NC(=O)C1=CN(C=CC1)[C@@H]2O[C@H](COP(=O)([O-])OP(=O)([O-])OC[C@H]3O[C@H]([C@H](OP(=O)([O-])[O-])[C@@H]3O)n4cnc5c(N)ncnc45)[C@@H](O)[C@H]2O
IUPAC Name tetrasodium;[(2R,3R,4R,5R)-2-(6-aminopurin-9-yl)-5-[[[[(2R,3S,4R,5R)-5-(3-carbamoyl-4H-pyridin-1-yl)-3,4-dihydroxy-tetrahydrofuran-2-yl]methoxy-oxido-phosphoryl]oxy-oxido-phosphoryl]oxymethyl]-4-hydroxy-tetrahydrofuran-3-yl] phosphate;hydrate
Molecular Weight (g/mol) 851.36
Formula Weight 851.0294
Chemical Name or Material beta-NADPH Tetrasodium Salt Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.