missing translation for 'onlineSavingsMsg'
Learn More

beta-Naphthoflavone, 98+%

Product Code. 11468467
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11468467 5 g 5g
11458467 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11468467 Supplier Thermo Scientific Alfa Aesar Supplier No. A18543.06
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6051-87-2 | C19H12O2 | 272.303 g/mol

Inhibitor of aryl hydrocarbon hydroxylase

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 6051-87-2
Molecular Formula C19H12O2
Molecular Weight (g/mol) 272.303
MDL Number MFCD00004986
InChI Key OUGIDAPQYNCXRA-UHFFFAOYSA-N
Synonym beta-naphthoflavone, 5,6-benzoflavone, beta-nf, 3-phenyl-1h-naphtho 2,1-b pyran-1-one, 3-phenyl-1h-benzo f chromen-1-one, 1h-naphtho 2,1-b pyran-1-one, 3-phenyl, unii-1bt0256y8o, 3-phenylbenzo f chromen-1-one, ccris 3262, .beta.-naphthoflavone
PubChem CID 2361
ChEBI CHEBI:77013
IUPAC Name 3-phenylbenzo[f]chromen-1-one
SMILES C1=CC=C(C=C1)C2=CC(=O)C3=C(O2)C=CC4=CC=CC=C43

Specifica

Melting Point 162°C to 166°C
Odor Odorless
Quantity 5 g
Beilstein 18991
Formula Weight 272.31
Percent Purity ≥98%
Chemical Name or Material beta-Naphthoflavone

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.