missing translation for 'onlineSavingsMsg'
Learn More

beta-Naphthoflavone, 99+%

Produktkod. 10497320 Handla allt Thermo Scientific Chemicals Produkter
Change view
Klicka för att se tillgängliga alternativ
Quantity:
1 g
5 g
Packaging:
Glass bottle
Förpackningsstorlek:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Quantity Packaging Förpackningsstorlek
10497320 5 g Glass bottle 5g
10561371 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 10497320 Leverantör Thermo Scientific Acros Leverantörsnummer 128170050
 Restricted Item
In Stock
Lägg i varukorgen
Lägg i varukorgen
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

CAS: 6051-87-2 | C19H12O2 | 272.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 6051-87-2
Molecular Formula C19H12O2
Molecular Weight (g/mol) 272.29
MDL Number MFCD00004986
InChI Key OUGIDAPQYNCXRA-UHFFFAOYSA-N
Synonym beta-naphthoflavone, 5,6-benzoflavone, beta-nf, 3-phenyl-1h-naphtho 2,1-b pyran-1-one, 3-phenyl-1h-benzo f chromen-1-one, 1h-naphtho 2,1-b pyran-1-one, 3-phenyl, unii-1bt0256y8o, 3-phenylbenzo f chromen-1-one, ccris 3262, .beta.-naphthoflavone
PubChem CID 2361
ChEBI CHEBI:77013
IUPAC Name 3-phenylbenzo[f]chromen-1-one
SMILES C1=CC=C(C=C1)C2=CC(=O)C3=C(O2)C=CC4=CC=CC=C43

Specifikationer

CAS Min % 99.0
CAS Max % 100.0
Melting Point 162.0°C to 164.0°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 99% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Beilstein 17, I, 216
Solubility Information Solubility in water: insoluble. Other solubilities: slightly soluble in ethanol,chloroform,soluble in sulfuric acid
Formula Weight 272.29
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material β-Naphthoflavone, 99+%
Visa mer Visa mindre

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.