missing translation for 'onlineSavingsMsg'
Learn More

Beta-Nicotinamide Adenine Dinucleotide, TRC

Product Code. 30007580
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007580 5 g 5g
30009121 250 mg 250mg
30009734 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007580 Supplier Toronto Research Chemicals Supplier No. TRCN4077845G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53-84-9
Molecular Formula C21 H27 N7 O14 P2
Molecular Weight (g/mol) 663.42
Synonym Adenosine 5'-(trihydrogen diphosphate), P'-5'-ester with 3-(aminocarbonyl)-1-β-D-ribofuranosylpyridinium hydroxide, inner salt, β-NAD, Codehydrase I, Coenzyme I, Enzopride, Nadide, Nicotinamide-adenine dinucleotide
IUPAC Name [[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxy-tetrahydrofuran-2-yl]methoxy-hydroxy-phosphoryl] [(2R,3S,4R,5R)-5-(3-carbamoylpyridin-1-ium-1-yl)-3,4-dihydroxy-tetrahydrofuran-2-yl]methyl phosphate
SMILES NC(=O)c1ccc[n+](c1)[C@@H]2O[C@H](COP(=O)([O-])OP(=O)(O)OC[C@H]3O[C@H]([C@H](O)[C@@H]3O)n4cnc5c(N)ncnc45)[C@@H](O)[C@H]2O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C21H27N7O14P2/c22-17-12-19(25-7-24-17)28(8-26-12)21-16(32)14(30)11(41-21)6-39-44(36,37)42-43(34,35)38-5-10-13(29)15(31)20(40-10)27-3-1-2-9(4-27)18(23)33/h1-4,7-8,10-11,13-16,20-21,29-32H,5-6H2,(H5-,22,23,24,25,33,34,35,36,37)/t10-,11-,13-,14-,15-,16-,20-,21-/m1/s1
Formula Weight 663.1091
Percent Purity >95%
Chemical Name or Material beta-Nicotinamide adenine dinucleotide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.