missing translation for 'onlineSavingsMsg'
Learn More

Beta-Nicotinamide Mononucleotide, TRC

Product Code. 17993185
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17993185 100 mg 100mg
17974745 10 mg 10mg
17942685 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17993185 Supplier Toronto Research Chemicals Supplier No. TRCN407765100MG
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1094-61-7
Molecular Formula C11 H15 N2 O8 P
Molecular Weight (g/mol) 334.22
Synonym Pyridinium, 3-(aminocarbonyl)-1-(5-O-phosphono-β-D-ribofuranosyl)-, inner salt, 3-Carbamoyl-1-β-D-ribofuranosylpyridinium hydroxide, 5'-phosphate, inner salt (6CI,7CI), Pyridinium, 3-(aminocarbonyl)-1-(5-O-phosphono-β-D-ribofuranosyl)-, hydroxide, inner salt, Pyridinium, 3-carbamoyl-1-β-D-ribofuranosyl-, hydroxide, 5'-(dihydrogen phosphate), inner salt (8CI), NMN, NMN (mononucleotide), Nicotinamide mononucleotide, Nicotinamide ribonucleoside 5'-phosphate, Nicotinamide ribonucleotide, Nicotinamide ribotide, β-D-NMN, β-NMN, β-Nicotinamide mononucleotide
IUPAC Name [(2R,3S,4R,5R)-5-(3-carbamoylpyridin-1-ium-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl hydrogen phosphate
SMILES NC(=O)c1ccc[n+](c1)[C@@H]2O[C@H](COP(=O)(O)[O-])[C@@H](O)[C@H]2O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C11H15N2O8P/c12-10(16)6-2-1-3-13(4-6)11-9(15)8(14)7(21-11)5-20-22(17,18)19/h1-4,7-9,11,14-15H,5H2,(H3-,12,16,17,18,19)/t7-,8-,9-,11-/m1/s1
Formula Weight 334.0566
Percent Purity >95%
Chemical Name or Material beta-Nicotinamide mononucleotide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.