missing translation for 'onlineSavingsMsg'
Learn More

beta-Propiolactone, 97%

Product Code. 11440797
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11440797 1 g 1g
11450797 5 g 5g
11460797 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11440797 Supplier Thermo Scientific Alfa Aesar Supplier No. B23197.03
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 57-57-8 | C3H4O2 | 72.06 g/mol

Beta-Propiolactone is widely used as a sterilizing and sporicidal agent, which is used to sterilize blood plasma, vaccines, tissue grafts, surgical instruments and enzymes. It finds an application in the manufacturing of acrylic acid and its esters. It is utilized as an intermediate in the synthesis of synthetic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Beta-Propiolactone is widely used as a sterilizing and sporicidal agent, which is used to sterilize blood plasma, vaccines, tissue grafts, surgical instruments and enzymes. It finds an application in the manufacturing of acrylic acid and its esters. It is utilized as an intermediate in the synthesis of synthetic compounds.

Solubility
Miscible with water and organic solvents.

Notes
Flammable material. Should be stored in a separate safety storage cabinet or room. Keep away from heat and sources of ignition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-57-8
Molecular Formula C3H4O2
Molecular Weight (g/mol) 72.06
MDL Number MFCD00005169
InChI Key VEZXCJBBBCKRPI-UHFFFAOYSA-N
Synonym beta-propiolactone, propiolactone, 2-oxetanone, propanolide, betaprone, 3-propanolide, 1,3-propiolactone, 3-propiolactone, beta-propionolactone, beta-propiolakton
PubChem CID 2365
ChEBI CHEBI:49073
IUPAC Name oxetan-2-one
SMILES O=C1CCO1

Specifications

Melting Point -34°C
Density 1.146
Boiling Point 161°C to 163°C
Flash Point 70°C (158°F)
Assay Percent Range 0.97
Refractive Index 1.413
Quantity 1 g
UN Number UN3382
Sensitivity Moisture sensitive
Merck Index 14,7820
Solubility Information Miscible with water and organic solvents.
Formula Weight 72.06
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material beta-Propiolactone
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • beta-Propiolactone
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H330-Fatal if inhaled.
  • H350-May cause cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands, and anyexposed skin thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P370+P378-In case of fire: Use dry sand, dry chemical, or alcohol-resistant foam to extinguish.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: .beta.-Propiolactone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.