missing translation for 'onlineSavingsMsg'
Learn More

beta-Sitosterol, with ca. 10% campesterol, ca. 75% beta-sitosterol

Product Code. 10664992 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
Unit Size:
100g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10664992 100 g 100g
10654992 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10664992 Supplier Thermo Scientific Acros Supplier No. 132721000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 83-46-5 | C29H50O | 414.72 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83-46-5
Molecular Formula C29H50O
InChI Key KZJWDPNRJALLNS-VJSFXXLFSA-N
Synonym beta-sitosterol, quebrachol, 22,23-dihydrostigmasterol, azuprostat, triastonal, harzol, cinchol, cupreol, rhamnol, sitosterol
PubChem CID 222284
ChEBI CHEBI:27693

Specifications

Color White
Loss on Drying 3.5% max.
Infrared Spectrum Authentic
Packaging Plastic Bottle
Quantity 100 g
Specific Rotation -24° to -32° (20°C, 589 nm) (c=2, CHCl3) Typically
SMILES CC[C@H](CC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C)C(C)C
IUPAC Name (1R,3aS,3bS,7S,9aR,9bS,11aR)-1-[(2R,5R)-5-ethyl-6-methylheptan-2-yl]-9a,11a-dimethyl-1H,2H,3H,3aH,3bH,4H,6H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-ol
Molecular Weight (g/mol) 414.72
Formula Weight 414.70
Percent Purity ≥75%
Physical Form Powder
Chemical Name or Material β-Sitosterol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.