missing translation for 'onlineSavingsMsg'
Learn More

Beta-Sitosterol (purity>80%), MedChemExpress

Product Code. 16462509
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16462509 1 g 1g
16459016 100 mg 100mg
16431328 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16462509 Supplier Medchem Express Supplier No. HYN0171
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Beta-Sitosterol (purity>80%) includes β-sitosterol (≥80%), stigmasterol, campesterol and brassicasterol mainly. Beta-Sitosterol is a plant sterol. Beta-Sitosterol (purity>80%) interfere with multiple cell signaling pathways, including cell cycle, apoptosis, proliferation, survival, invasion, angiogenesis, metastasis and inflammation.

Beta-Sitosterol (purity>80%) includes β-sitosterol (≥80%), stigmasterol, campesterol and brassicasterol mainly. Beta-Sitosterol is a plant sterol. Beta-Sitosterol (purity>80%) interfere with multiple cell signaling pathways, including cell cycle, apoptosis, proliferation, survival, invasion, angiogenesis, metastasis and inflammation.

Specifications

Chemical Name or Material Beta-Sitosterol (purity>80%)
CAS 83-46-5
Color White
Purity Grade Notes Research
Molecular Formula C29H50O
Quantity 1 g
Synonym β-Sitosterol (purity>80%) 22,23-Dihydrostigmasterol (purity>80%)
Solubility Information Ethanol : 3.85 mg/mL (9.28 mM; Need ultrasonic) ∣DMSO : 1 mg/mL (2.41 mM; Need ultrasonic) ∣H2O : 0.1 mg/mL (0.24 mM; Need ultrasonic)
SMILES CC[C@@H](C(C)C)CC[C@@H](C)[C@H]1CC[C@@]2([H])[C@]3([H])CC=C4C[C@@H](O)CC[C@]4(C)[C@@]3([H])CC[C@]12C.[(purity>80%)]
Molecular Weight (g/mol) 414.71
Formula Weight 414.71
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.