missing translation for 'onlineSavingsMsg'
Learn More

Beta-Sitosterol (Synthetic), TRC

Product Code. 17999344
Change view
Click to view available options
Quantity:
1 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17999344 10 mg 10mg
17998984 1 mg 1mg
17988504 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17999344 Supplier Toronto Research Chemicals Supplier No. TRCS49705010MG
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83-46-5
Molecular Formula C29 H50 O
Molecular Weight (g/mol) 414.71
Synonym Nimbosterol, Stigmast-5-en-3beta-ol, (-)-beta-Sitosterol, (24R)-Ethylcholest-5-en-3beta-ol, (24R)-Stigmast-5-en-3beta-ol, 22,23-Dihydrostigmasterol, 24alpha-Ethylcholesterol, Angelicin, Azuprostat, Betaprost, Cinchol, Cupreol, Harzol, Inmunicin MAYMO, NSC 18173, NSC 49083, NSC 8096, Prostasal, Quebrachol, Rhammol, Rhamnol, SKF 14463, Sito-Lande, Sitosterol, Sobatum, Stigmasta-5-en-3beta-ol, Stigmasterol, 22,23-dihydro-, delta5-Stigmasten-3beta-ol, alpha-Dihydrofucosterol, alpha-Phytosterol, beta-Sitosterin, beta-Sitosterol
IUPAC Name (3S,8S,9S,10R,13R,14S,17R)-17-[(2R,5R)-5-ethyl-6-methylheptan-2-yl]-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-ol
SMILES CC[C@H](CC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C)C(C)C

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C29H50O/c1-7-21(19(2)3)9-8-20(4)25-12-13-26-24-11-10-22-18-23(30)14-16-28(22,5)27(24)15-17-29(25,26)6/h10,19-21,23-27,30H,7-9,11-18H2,1-6H3/t20-,21-,23+,24+,25-,26+,27+,28+,29-/m1/s1
Formula Weight 414.3862
Percent Purity >95%
Chemical Name or Material beta-Sitosterol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.