missing translation for 'onlineSavingsMsg'
Learn More

Betahistine Dihydrochloride, TRC

Product Code. 17972555
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17972555 1 g 1g
17934265 5 g 5g
17963835 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17972555 Supplier Toronto Research Chemicals Supplier No. TRCB3240011G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5579-84-0
Molecular Formula C8 H12 N2 . 2 Cl H
Molecular Weight (g/mol) 209.12
Synonym Betahistine Dihydrochloride, N-Methyl-2-pyridineethanamine hydrochloride (1:2), N-Methyl-2-pyridineethanamine dihydrochloride, 2-[2-(Methylamino)ethyl]pyridine dihydrochloride, Betahistine dihydrochloride, Betahistine hydrochloride, Betaserc, PT 9, Serc, Sinmenier, Vasomotal
IUPAC Name N-methyl-2-pyridin-2-ylethanamine;dihydrochloride
SMILES Cl.Cl.CNCCc1ccccn1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C8H12N2.2ClH/c1-9-7-5-8-4-2-3-6-10-8;;/h2-4,6,9H,5,7H2,1H3;2*1H
Formula Weight 208.0534
Percent Purity >95%
Chemical Name or Material Betahistine dihydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.