missing translation for 'onlineSavingsMsg'
Learn More

Betaine, 98%, for analysis, anhydrous

Product Code. 13468448 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
100g
2.5kg
500g
5g
Product Code. Quantity Packaging unitSize
13468448 2.5 kg Plastic bottle 2.5kg
10246930 5 g Glass Bottle 5g
10591451 100 g Plastic bottle 100g
10721424 500 g Plastic bottle 500g
4 options
This item is not returnable. View return policy

Product Code. 13468448

Brand: Thermo Scientific Acros 204240025

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 107-43-7 | C5H11NO2 | 117.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-43-7
Molecular Formula C5H11NO2
Molecular Weight (g/mol) 117.15
MDL Number MFCD00012123
InChI Key KWIUHFFTVRNATP-UHFFFAOYSA-N
Synonym betaine, glycine betaine, oxyneurine, lycine, trimethylglycine, abromine, trimethylglycocoll, glycylbetaine, glycocoll betaine, acidin-pepsin
PubChem CID 247
ChEBI CHEBI:17750
IUPAC Name 2-(trimethylazaniumyl)acetate
SMILES C[N+](C)(C)CC(=O)[O-]

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 301.0°C
Color White
Loss on Drying 1% max. (140°C, 3 hrs)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Linear Formula (CH3)3NCH2CO2
Quantity 2.5 kg
Beilstein 04, IV, 2369
Merck Index 15, 1181
Solubility Information Solubility in water: 1000g/L (25°C). Other solubilities: 55g/100 g methanol,8.7g/100 g ethanol,sparingly soluble in ether,slightly soluble in chloroform
Transmittance 95% min. (c=10, H2O, 430nm) 1cm cell
Formula Weight 117.15
Percent Purity 98%
Grade Analysis
Physical Form Crystalline Powder or Crystals
Chemical Name or Material βine, Anhydrous
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.