missing translation for 'onlineSavingsMsg'
Learn More

Betamethasone 17-Valerate, TRC

Product Code. 30337935
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30337935 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30337935 Supplier Toronto Research Chemicals Supplier No. TRCB32707550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Betamethasone Valerate
CAS 2152-44-5
Purity Grade Notes HPLC
Molecular Formula C27 H37 F O6
Quantity 50 mg
Synonym Pregna-1,4-diene-3,20-dione, 9-fluoro-11β,17,21-trihydroxy-16β-methyl-, 17-valerate (7CI,8CI), (11β,16β)-9-Fluoro-11,21-dihydroxy-16-methyl-17-[(1-oxopentyl)oxy]pregna-1,4-diene-3,20-dione, Valeric acid, 17-ester with 9-fluoro-11β,17,21-trihydroxy-16β-methylpregna-1,4-diene-3,20-dione (8CI), 9α-Fluoro-11β,21-dihydroxy-16β-methyl-17α-valeryloxypregna-1,4-diene-3,20-dione, 9α-Fluoro-16β-methylprednisolone 17-valerate, Bedermin, Beta-Val, Betamethasone 17-valerate, Betamethasone 17α-valerate, Betamethasone valerate, Betnesol V, Betneval, Betnovate, Betnovateat, Betoid, Bettamousse, Bextasol, Celestan V, Celestane V, Celestoderm, Celestoderm V, Celeston valerate, Dermosol, Dermovaleas, Ecoval 70, Fuciderm, Hormezon, Rinderon V, Stanoval, Tokuderm, Valisone, β-Methasone 17-valerate
InChI Formula InChI=1S/C27H37FO6/c1-5-6-7-23(33)34-27(22(32)15-29)16(2)12-20-19-9-8-17-13-18(30)10-11-24(17,3)26(19,28)21(31)14-25(20,27)4/h10-11,13,16,19-21,29,31H,5-9,12,14-15H2,1-4H3/t16-,19-,20-,21-,24-,25-,26-,27-/m0/s1
SMILES CCCCC(=O)O[C@@]1([C@@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C)C(=O)CO
IUPAC Name [(8S,9R,10S,11S,13S,14S,16S,17R)-9-fluoro-11-hydroxy-17-(2-hydroxyacetyl)-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl] pentanoate
Molecular Weight (g/mol) 476.58
Formula Weight 476.2574
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.