missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Betamethasone 17-Valerate, TRC
Specifications
Specifications
| Chemical Name or Material | Betamethasone Valerate |
| CAS | 2152-44-5 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C27 H37 F O6 |
| Quantity | 1 g |
| Synonym | Pregna-1,4-diene-3,20-dione, 9-fluoro-11β,17,21-trihydroxy-16β-methyl-, 17-valerate (7CI,8CI), (11β,16β)-9-Fluoro-11,21-dihydroxy-16-methyl-17-[(1-oxopentyl)oxy]pregna-1,4-diene-3,20-dione, Valeric acid, 17-ester with 9-fluoro-11β,17,21-trihydroxy-16β-methylpregna-1,4-diene-3,20-dione (8CI), 9α-Fluoro-11β,21-dihydroxy-16β-methyl-17α-valeryloxypregna-1,4-diene-3,20-dione, 9α-Fluoro-16β-methylprednisolone 17-valerate, Bedermin, Beta-Val, Betamethasone 17-valerate, Betamethasone 17α-valerate, Betamethasone valerate, Betnesol V, Betneval, Betnovate, Betnovateat, Betoid, Bettamousse, Bextasol, Celestan V, Celestane V, Celestoderm, Celestoderm V, Celeston valerate, Dermosol, Dermovaleas, Ecoval 70, Fuciderm, Hormezon, Rinderon V, Stanoval, Tokuderm, Valisone, β-Methasone 17-valerate |
| InChI Formula | InChI=1S/C27H37FO6/c1-5-6-7-23(33)34-27(22(32)15-29)16(2)12-20-19-9-8-17-13-18(30)10-11-24(17,3)26(19,28)21(31)14-25(20,27)4/h10-11,13,16,19-21,29,31H,5-9,12,14-15H2,1-4H3/t16-,19-,20-,21-,24-,25-,26-,27-/m0/s1 |
| SMILES | CCCCC(=O)O[C@@]1([C@@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C)C(=O)CO |
| IUPAC Name | [(8S,9R,10S,11S,13S,14S,16S,17R)-9-fluoro-11-hydroxy-17-(2-hydroxyacetyl)-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl] pentanoate |
| Molecular Weight (g/mol) | 476.58 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?