missing translation for 'onlineSavingsMsg'
Learn More

Betamethasone 17-Valerate, TRC

Product Code. 30331773
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30331773 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30331773 Supplier Toronto Research Chemicals Supplier No. TRCB32707525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Especificaciones

Chemical Name or Material Betamethasone Valerate
CAS 2152-44-5
Purity Grade Notes HPLC
Molecular Formula C27 H37 F O6
Quantity 25 mg
Synonym Pregna-1,4-diene-3,20-dione, 9-fluoro-11β,17,21-trihydroxy-16β-methyl-, 17-valerate (7CI,8CI), (11β,16β)-9-Fluoro-11,21-dihydroxy-16-methyl-17-[(1-oxopentyl)oxy]pregna-1,4-diene-3,20-dione, Valeric acid, 17-ester with 9-fluoro-11β,17,21-trihydroxy-16β-methylpregna-1,4-diene-3,20-dione (8CI), 9α-Fluoro-11β,21-dihydroxy-16β-methyl-17α-valeryloxypregna-1,4-diene-3,20-dione, 9α-Fluoro-16β-methylprednisolone 17-valerate, Bedermin, Beta-Val, Betamethasone 17-valerate, Betamethasone 17α-valerate, Betamethasone valerate, Betnesol V, Betneval, Betnovate, Betnovateat, Betoid, Bettamousse, Bextasol, Celestan V, Celestane V, Celestoderm, Celestoderm V, Celeston valerate, Dermosol, Dermovaleas, Ecoval 70, Fuciderm, Hormezon, Rinderon V, Stanoval, Tokuderm, Valisone, β-Methasone 17-valerate
InChI Formula InChI=1S/C27H37FO6/c1-5-6-7-23(33)34-27(22(32)15-29)16(2)12-20-19-9-8-17-13-18(30)10-11-24(17,3)26(19,28)21(31)14-25(20,27)4/h10-11,13,16,19-21,29,31H,5-9,12,14-15H2,1-4H3/t16-,19-,20-,21-,24-,25-,26-,27-/m0/s1
SMILES CCCCC(=O)O[C@@]1([C@@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C)C(=O)CO
IUPAC Name [(8S,9R,10S,11S,13S,14S,16S,17R)-9-fluoro-11-hydroxy-17-(2-hydroxyacetyl)-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl] pentanoate
Molecular Weight (g/mol) 476.58
Formula Weight 476.2574
Percent Purity >95
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.