missing translation for 'onlineSavingsMsg'
Learn More

Betamethasone 21-Phosphate Disodium Salt, TRC

Product Code. 30333019
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
This item is not returnable. View return policy

Product Code. 30333019

Brand: Toronto Research Chemicals TRCB327030250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Betamethasone Sodium Phosphate
CAS 151-73-5
Purity Grade Notes HPLC
Molecular Formula C22 H28 F O8 P . 2 Na
Quantity 250 mg
Synonym Betamethasone Sodium Phosphate, Dexamethasone Sodium Phosphate Imp. B (EP), (11β,16β)-9-Fluoro-11,17-dihydroxy-16-methyl-21-(phosphonooxy)pregna-1,4-diene-3,20-dione sodium salt (1:2), (11β,16β)-9-Fluoro-11,17-dihydroxy-16-methyl-21-(phosphonooxy)pregna-1,4-diene-3,20-dione disodium salt, 9-Fluoro-11β,17,21-trihydroxy-16β-methylpregna-1,4-diene-3,20-dione 21-(dihydrogen phosphate) disodium salt, Bentelan, Betamethasone 21-disodium phosphate, Betamethasone 21-phosphate disodium salt, Betamethasone disodium phosphate, Betamethasone sodium phosphate, Betnesol, Celestan solubile, Disodium betamethasone 21-phosphate, Durabetason, NSC 90616, Steronema, Vista-Methasone
InChI Formula InChI=1S/C22H30FO8P.2Na/c1-12-8-16-15-5-4-13-9-14(24)6-7-19(13,2)21(15,23)17(25)10-20(16,3)22(12,27)18(26)11-31-32(28,29)30;;/h6-7,9,12,15-17,25,27H,4-5,8,10-11H2,1-3H3,(H2,28,29,30);;/q;2*+1/p-2/t12-,15-,16-,17-,19-,20-,21-,22-;;/m0../s1
SMILES [Na+].[Na+].C[C@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)COP(=O)([O-])[O-]
IUPAC Name disodium;[2-[(8S,9R,10S,11S,13S,14S,16S,17R)-9-fluoro-11,17-dihydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] phosphate
Molecular Weight (g/mol) 516.4
Formula Weight 516.1301
Percent Purity >95
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.