missing translation for 'onlineSavingsMsg'
Learn More

Betamethasone, MedChemExpress

Product Code. 16486226
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16486226 100 mg 100mg
16421676 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16486226 Supplier Medchem Express Supplier No. HY13570
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Betamethasone is a synthetic glucocorticoid with anti-inflammatory and immunosuppressive activities. Betamethasone accelerates fetal lung maturation and induces gene expression and apoptosis.

Betamethasone is a synthetic glucocorticoid with anti-inflammatory and immunosuppressive activities. Betamethasone accelerates fetal lung maturation and induces gene expression and apoptosis.

Specifications

Chemical Name or Material Betamethasone
CAS 378-44-9
Color White
Purity Grade Notes Research
Molecular Formula C22H29FO5
Quantity 100 mg
Solubility Information DMSO : ≥ 50 mg/mL (127.40 mM)
SMILES C[C@@]12[C@](C[C@H](C)[C@]2(O)C(CO)=O)([H])[C@]3([H])CCC4=CC(C=C[C@]4(C)[C@@]3(F)[C@@H](O)C1)=O
Molecular Weight (g/mol) 392.46
Formula Weight 392.46
Percent Purity 99.91%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.