missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Betamethasone valerate

Product Code. 16594294 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16594294 1 g 1g
16504304 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16594294 Supplier Thermo Scientific Chemicals Supplier No. 464930010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2152-44-5
Molecular Formula C27H37FO6
Molecular Weight (g/mol) 476.59
InChI Key SNHRLVCMMWUAJD-SUYDQAKGSA-N
IUPAC Name (1R,2S,3aS,3bS,9aS,9bR,10S,11aS)-9b-fluoro-10-hydroxy-1-(2-hydroxyacetyl)-2,9a,11a-trimethyl-7-oxo-1H,2H,3H,3aH,3bH,4H,5H,7H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl pentanoate
SMILES CCCCC(=O)O[C@@]1([C@@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C)C(=O)CO

Specifications

Quantity 1 g
Formula Weight 476.58
Chemical Name or Material βmethasone valerate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.