missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ BETP
GREENER_CHOICE

Product Code. 30001983
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 30001983

Brand: Tocris Bioscience™ 4778/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Positive allosteric modulator of GLP-1 receptors

Selective positive allosteric modulator and partial agonist of the glucagon-like peptide 1 (GLP-1) receptor. Increases binding affinity of oxyntomodulin for the GLP-1 receptor. Potentiates oxyntomodulin-mediated GLP-1 receptor signaling in vitro and insulin secretion in vivo. Has no effect on GLP-2, GIP, PTH or glucagon receptors.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1371569-69-5
Molecular Formula C20H17F3N2O2S

Specifications

Quantity 10 mg
Formula Weight 406.42
Molecular Weight (g/mol) 406.423
Chemical Name or Material BETP
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.