missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Betulinic acid

Product Code. 18084452
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18084452 10 mg 10mg
18034982 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18084452 Supplier R&D Systems™ Supplier No. 3906/10
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Antitumor and anti-HIV agent; activates NF-κB. Also TGR5 agonist

Betulinic acid is a natural triterpenoid that displays anti-HIV and antitumor activity. Induces the production of reactive oxygen species (ROS) and activates NF-κB. Also a GPBA receptor partial agonist (EC50 = 1.04 μM, efficacy 83%). Betulinic acid suppresses areobic glycolysis by regulating the Cav-1/NF-κB/c-Myc pathway in and inhibits growth in breast cancer cells.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 472-15-1
Molecular Formula C30H48O3

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 456.7
Purity >98%
Molecular Weight (g/mol) 456.711
Chemical Name or Material Betulinic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.