missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Betulinic acid

Product Code. 15881012 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15881012 250 mg 250mg
15871012 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15881012 Supplier Thermo Scientific Chemicals Supplier No. 462872500
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 472-15-1 | C30H48O3 | 456.71 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 472-15-1
Molecular Formula C30H48O3
Molecular Weight (g/mol) 456.71
InChI Key QGJZLNKBHJESQX-FZFNOLFKSA-N
PubChem CID 64971
ChEBI CHEBI:3087

Specifications

Quantity 250 mg
Chemical Name or Material Betulinic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.