missing translation for 'onlineSavingsMsg'
Learn More

Betulinic acid, MedChemExpress

Product Code. 16481576
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16481576 500 mg 500mg
16465016 100 mg 100mg
16420996 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16481576 Supplier Medchem Express Supplier No. HY10529
 Restricted Item
Estimated Shipment: 04-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Betulinic acid is a natural pentacyclic triterpenoid, acts as a eukaryotic topoisomerase I inhibitor, with an IC50 of 5 μM, and possesses anti-HIV, anti-malarial, anti-inflammatory and anti-tumor properties.

Betulinic acid is a natural pentacyclic triterpenoid, acts as a eukaryotic topoisomerase I inhibitor, with an IC50 of 5 μM, and possesses anti-HIV, anti-malarial, anti-inflammatory and anti-tumor properties.

Specifications

Chemical Name or Material Betulinic acid
CAS 472-15-1
Color Off-White
Purity Grade Notes Research
Molecular Formula C30H48O3
Quantity 500 mg
Synonym Lupatic acid Betulic acid
Solubility Information DMSO : 35.71 mg/mL (78.19 mM; Need ultrasonic)
SMILES [H][C@@]12CC[C@@]3(C)[C@]4(C)CC[C@]5(C(O)=O)[C@@]([C@H](C(C)=C)CC5)([H])[C@@](CC[C@@]([H])3[C@@]1(C)CC[C@H](O)C2(C)C)4[H]
Molecular Weight (g/mol) 456.7
Formula Weight 456.7
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.