missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MedChemExpress Betulonic acid
Specifications
Specifications
| Content And Storage | Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month |
| Molecular Weight (g/mol) | 454.68 g/mol |
| Shipping Condition | Room temperature |
| For Use With (Application) | Cancer-programmed cell death |
| CAS | 4481-62-3 |
| Product Type | Natural Products |
| Purity | 98.0% |
| Solubility | DMSO : 6.67 mg/mL (14.67 mM∣Need ultrasonic) |
| Synonym | Betunolic acid∣ Liquidambaric acid∣ (+)-Betulonic acid |
| Target | Parasite∣HSV |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?