missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Bexarotene

Product Code. 18768788
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18768788 50 mg 50mg
18731348 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18768788 Supplier R&D Systems™ Supplier No. 5819/50
  • ONLINE EXCLUSIVE PRICE
    447.09 €
    402.15 € / 50mg
    Save 44.94 €
    10% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent and selective RXR agonist

Bexarotene is a potent and selective RXR agonist (EC50 values are 24, 25 and 33 nM for RXRβ, RXRγ and RXRα, respectively). Exhibits >300-fold selectivity for RXR over RAR receptors. Inhibits tumor cell invasion and migration in vitro, and inhibits metastasis and angiogenesis in mouse models. Also promotes brown adipogenic differentiation and induces brown adipogenic reprogramming from myoblasts.
TRUSTED_SUSTAINABILITY

Spezifikation

Chemical Name or Material 4-[1-(5,6,7,8-Tetrahydro-3,5,5,8,8-pentamethyl-2-naphthalenyl)ethenyl]benzoic acid
CAS 153559-49-0
Synonym LGD 1069
Quantity 50 mg
Product Line Tocris
Target Retinoid X Receptor Agonists
Molecular Formula C24H28O2
Formula Weight 348.48
Purity >99%
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.