missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ BF 738735

Product Code. 18767836
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy

Product Code. 18767836

Brand: Tocris Bioscience™ 6246/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective PI 4-kinase IIIβ inhibitor; broad spectrum enterovirus replication inhibitor

BF 738735 is a potent and selective ATP-competitive phosphatidylinositol 4-kinase III beta (PI 4-KIIIβ) inhibitor (IC50 = 5.7 nM). Displays approximately 200-fold selectivity over PI 4-KIIIα. Broad spectrum enterovirus replication inhibitor.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Fluoro-4-[2-methyl-8-[[[3-(methylsulfonyl)phenyl]methyl]amino]imidazo[1,2-a]pyrazin-3-yl]phenol
CAS 1436383-95-7
Quantity 10 mg
Target Other Kinase Inhibitors
Molecular Formula C21H19FN4O3S
Formula Weight 426.47
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.