Learn More
Scientific Industries BG Horz 12 Tube Holder, 1.5 mL or 2.0 mL
Product Code. 17201752 Shop All Ohaus Products
Change view
Click to view available options
For Use With (Equipment):
Bead Genie
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
17201752 Bead Genie Each
1 options
Product Code. 17201752 Supplier Scientific Industries Supplier No. PPB100404
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
BG Horz 12 Tube Holder, 1.5 mL or 2.0 mL is an accessory for the Bead Genie consisting of a holder for up to 12 tubes at once
  • Requires clamping disk supplied with unit
TRUSTED_SUSTAINABILITY
Specifications
Product Type Tube Holder
For Use With (Application) Bead Beating
For Use With (Equipment) Bead Genie
Holds 12 x 1.5 or 2.0 mL Tubes
Model PP-B100-404
Packaging Quantity 1 Ea.