Learn More
Scientific Industries BG Vert Holder
Product Code. 17291742 Shop All Ohaus Products
Change view
Click to view available options
Holds:
12 x 1.5 or 2.0 mL Tubes
3 x 50 mL Tubes
6 x 15 mL Tubes
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds Unit Size
17291742 3 x 50 mL Tubes Each
17271742 12 x 1.5 or 2.0 mL Tubes Each
17281742 6 x 15 mL Tubes Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17291742 Supplier Scientific Industries Supplier No. PPB100403
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
BG Vert Holder is a tube holder accessory available in a variety of sizes for use with the Bead Genie.
  • Requires clamping disk supplied with the unit
TRUSTED_SUSTAINABILITY
Specifications
Product Type Vertical Holder
For Use With (Application) Bead Beating
For Use With (Equipment) Bead Genie
Holds 3 x 50 mL Tubes
Model PP-B100-403
Packaging Quantity 1 Ea.