Learn More
Keystone Cleanroom Products BHD Blue Autoclaveable Wrapper
When secured with autoclave tape or elasticized bands, Keystone Cleanroom Product’s Blue BHD wrappers provide an adequate microbial barrier from external sources of contamination.
Brand:  Keystone Cleanroom Products BHDSHEET4545
Additional Details : Weight : 24.49780kg
 View more versions of this product
Product Code. 17748197
  • 1215.00 € / Pack of 250
Estimated Shipment: 01-07-2024
to see stock.
Add to basket
Description
Description
  • Designed for Steam Sterilization
  • Treated to impart water and alcohol repellency
  • Wet formed, lightweight non-woven fabric comprised of natural wood pulp and polyester fibers bonded with a resin binder
  • Sufficient Microbial Barrier
  • Resists Tearing and Punctures
  • Does not contain any natural latex rubber
  • Processed and Packaged in ISO Certified Cleanrooms
  • Certificates of Conformance sent with shipment
Specifications
Specifications
45 x 45 in.