missing translation for 'onlineSavingsMsg'
Learn More

Abcam BHHCT, Fluorescent Sensitizer, Eu3+ Labelling, Powder, 5 mg

Product Code. 32117558
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32117558 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32117558 Supplier Abcam Supplier No. ab1453145mg
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Increases luminescence intensity 11-fold and measures peroxidase activity and oxidative stress levels.

  • Fluorescent sensitizer for Eu3+ labelling
  • Increases luminescence intensity 11-fold
  • Measures peroxidase activity and oxidative stress levels
  • Persists as an Eu3+ complex for 500 μs
  • Molecular weight: 804.9 Da
  • Purity: >90%
  • Form: Powder

Specifications

Buffer 100% BHHCT
Content And Storage Store at -20°C.
Form Powder
Reactivity Peroxidase
Shipping Condition Ambient
Substrate Properties Chemical Substrate
Substrate Peroxidase
Target Molecule Eu3+
Detection Method Fluorescence
Formulation 100% BHHCT
Quantity 5 mg
For Use With (Application) Eu3+ labelling, peroxidase activity measurement, oxidative stress detection
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.